siRNA Details
| Gene Symbol: Capn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009796.2 | NP_033926.1 | 18, 19 | 2224 | ||
| GenBank mRNA | AB028640.1 | - | 18, 19 | 2224 | ||
| AJ012475.1 | - | 18, 19 | 2037 | |||
| AK154277.1 | - | 18, 19 | 2310 | |||
| AK169334.1 | - | 18, 19 | 3269 | |||
| BC095980.1 | - | 18, 19 | 3239 | |||

Capn7
Genomic Map