siRNA Details
| Gene Symbol: Cask | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284503.1 | NP_001271432.1 | 17 | 2124 | ||
| NM_001284504.1 | NP_001271433.1 | 16 | 2106 | |||
| NM_001284505.1 | NP_001271434.1 | 16 | 2106 | |||
| NM_009806.3 | NP_033936.2 | 16 | 2106 | |||
| GenBank mRNA | AK136523.1 | - | 9 | 802 | ||
| BC141292.1 | - | 16 | 1790 | 9055904 | ||
| BC145620.1 | - | 16 | 1790 | 9054188 | ||
| Y17137.1 | - | 17 | 2126 | |||
| Y17138.1 | - | 17 | 2126 | |||

Cask
Genomic Map