siRNA Details
| Gene Symbol: Runx1t1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111026.2 | NP_001104496.1 | 6 | 1243 | ||
| NM_001111027.2 | NP_001104497.1 | 7 | 1437 | |||
| NM_009822.3 | NP_033952.1 | 6 | 1175 | |||
| GenBank mRNA | AB221624.1 | - | 5 | 822 | ||
| AK032132.1 | - | 6 | 1110 | |||
| AK142006.1 | - | 6 | 1244 | |||
| BC139200.1 | - | 6 | 901 | 8862222 | ||
| BC139201.1 | - | 6 | 905 | 8862223 | ||
| D32007.1 | - | 6 | 1175 | |||
| X79989.1 | - | 6 | 882 | |||

Runx1t1
Genomic Map