siRNA Details
| Gene Symbol: Cbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007619.2 | NP_031645.2 | 14 | 2359 | ||
| GenBank mRNA | AK029826.1 | - | 14 | 2362 | ||
| AK153915.1 | - | 14 | 2311 | |||
| AK163907.1 | - | 13 | 2185 | |||
| BC125285.1 | - | 14 | 2274 | 40129800 | ||
| X57111.1 | - | 14 | 2246 | |||

Cbl
Genomic Map