siRNA Details
| Gene Symbol: Cbr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007621.2 | NP_031647.1 | 6 | 521 | ||
| GenBank mRNA | AK081816.1 | - | 6 | 492 | ||
| BC010758.1 | - | 6 | 481 | 4219349 | ||
| D26123.1 | - | 6 | 508 | |||
| X07411.1 | - | 6 | 478 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbr2| Gene Symbol: Cbr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007621.2 | NP_031647.1 | 6 | 521 | ||
| GenBank mRNA | AK081816.1 | - | 6 | 492 | ||
| BC010758.1 | - | 6 | 481 | 4219349 | ||
| D26123.1 | - | 6 | 508 | |||
| X07411.1 | - | 6 | 478 | |||