siRNA Details
| Gene Symbol: Cbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007622.3 | NP_031648.1 | 2 | 349 | ||
| GenBank mRNA | AK014215.1 | - | 2 | 325 | ||
| AK017608.1 | - | 2 | 238 | |||
| AK157710.1 | - | 2 | 351 | |||
| BC055290.1 | - | 2 | 292 | 6313666 | ||
| X56690.1 | - | 1 | 94 | |||

Cbx1
Genomic Map