siRNA Details
| Gene Symbol: Cbx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076789.1 | NP_001070257.1 | 3 | 319 | ||
| NM_001110216.1 | NP_001103686.1 | 4 | 420 | |||
| NM_007626.3 | NP_031652.1 | 3 | 320 | |||
| GenBank mRNA | AK008792.1 | - | 3 | 355 | ||
| AK018349.1 | - | 3 | 266 | |||
| AK030366.1 | - | 3 | 320 | |||
| AK030442.1 | - | 4 | 373 | |||
| AK032975.1 | - | 3 | 276 | |||
| BC004707.1 | - | 3 | 319 | 3499489 | ||
| X99641.1 | - | 3 | 252 | |||

Cbx5
Genomic Map