siRNA Details
| Gene Symbol: Ccnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007630.2 | NP_031656.2 | 3 | 346 | ||
| GenBank mRNA | AK013312.1 | - | 3 | 315 | ||
| AK076122.1 | - | 3 | 340 | |||
| AK178328.1 | - | 2 | 143 | |||
| BC008247.1 | - | 3 | 262 | 3156950 | ||
| BY300844.1 | - | 3 | 349 | |||
| CT010286.1 | - | 3 | 232 | |||
| X66032.1 | - | 3 | 333 | |||

Ccnb2
Genomic Map