siRNA Details
| Gene Symbol: Ccnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007630.2 | NP_031656.2 | 4 | 469 | ||
| GenBank mRNA | AK013312.1 | - | 4 | 438 | ||
| AK076122.1 | - | 4 | 463 | |||
| BC008247.1 | - | 4 | 385 | 3156950 | ||
| CT010286.1 | - | 4 | 355 | |||
| X66032.1 | - | 4 | 456 | |||

Ccnb2
Genomic Map