siRNA Details
| Gene Symbol: Ccnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007630.2 | NP_031656.2 | 2 | 237 | ||
| GenBank mRNA | AK013312.1 | - | 2 | 206 | ||
| AK076122.1 | - | 2 | 231 | |||
| BC008247.1 | - | 2 | 153 | 3156950 | ||
| BY300844.1 | - | 2 | 240 | |||
| CT010286.1 | - | 2 | 123 | |||
| X66032.1 | - | 2 | 224 | |||

Ccnb2
Genomic Map