siRNA Details
| Gene Symbol: GUCY2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004963.3 | NP_004954.2 | 12, 13 | 1618 | ||
| XM_011520631.2 | XP_011518933.1 | 12, 13 | 1516 | |||
| GenBank mRNA | BC136544.1 | - | 12, 13 | 1527 | 9020534 | |
| BC136545.1 | - | 12, 13 | 1527 | 9020535 | ||
| M73489.1 | - | 12, 13 | 1529 | |||
| S57551.1 | - | 12, 13 | 1598 | |||

GUCY2C
Genomic Map