siRNA Details
| Gene Symbol: Ccnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009833.1 | NP_033963.1 | 9 | 1361 | ||
| GenBank mRNA | AF087662.1 | - | 9 | 1361 | ||
| AF095640.1 | - | 9 | 1361 | |||
| AF109179.1 | - | 9 | 1361 | |||
| AF113951.1 | - | 9 | 1361 | |||
| AK133168.1 | - | 2061 | ||||
| BC131685.1 | - | 9 | 1361 | 40111509 | ||
| BC131686.1 | - | 9 | 1361 | |||

Ccnt1
Genomic Map