siRNA Details
| Gene Symbol: Cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013486.2 | NP_038514.1 | 3, 4 | 648 | ||
| GenBank mRNA | AF065906.1 | - | 3, 4 | 645 | ||
| AF065907.1 | - | 3, 4 | 645 | |||
| AF065908.1 | - | 3, 4 | 645 | |||
| AF065909.1 | - | 3, 4 | 645 | |||
| AF306543.1 | - | 3, 4 | 645 | |||
| BC053731.1 | - | 3, 4 | 631 | 30077134 | ||
| M18934.1 | - | 3, 4 | 635 | |||
| X06143.1 | - | 3, 4 | 603 | |||
| Y00023.1 | - | 3, 4 | 648 | |||

Cd2
Genomic Map