siRNA Details
| Gene Symbol: GYPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308187.1 | NP_001295116.1 | 3 | 334 | ||
| NM_001308190.1 | NP_001295119.1 | 2 | 235 | |||
| NM_002099.7 | NP_002090.4 | 3 | 334 | |||
| XM_017008134.1 | XP_016863623.1 | 3 | 338 | |||
| XM_017008135.1 | XP_016863624.1 | 4 | 401 | |||
| XM_017008136.1 | XP_016863625.1 | 2 | 303 | |||
| GenBank mRNA | AK290561.1 | - | 3 | 299 | ||
| AY950614.1 | - | 3 | 183 | |||
| BC005319.1 | - | 3 | 240 | 3930700 | ||
| BC013328.1 | - | 3 | 221 | 4134368 | ||
| DA537533.1 | - | 3 | 299 | |||
| DA539682.1 | - | 3 | 335 | |||
| EU338231.1 | - | 4 | 303 | |||
| EU338232.1 | - | 3 | 220 | |||
| EU338233.1 | - | 4 | 303 | |||
| EU338234.1 | - | 2 | 127 | |||
| GU347002.1 | - | 4 | 327 | |||
| GU347003.1 | - | 4 | 406 | |||
| GU347004.1 | - | 4 | 384 | |||
| GU347006.1 | - | 3 | 226 | |||
| GU347008.1 | - | 2 | 121 | |||
| L31860.1 | - | 3 | 238 | |||
| M12857.1 | - | 3 | 193 | |||
| M36281.1 | - | 3 | 193 | |||
| U00177.1 | - | 3 | 183 | |||
| X08054.1 | - | 3 | 239 | |||
| X51798.1 | - | 3 | 207 | |||
| X57023.1 | - | 3 | 183 | |||

GYPA
Genomic Map