siRNA Details
| Gene Symbol: Cd28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007642.4 | NP_031668.3 | 2 | 225 | ||
| GenBank mRNA | AK030812.1 | - | 2 | 147 | ||
| AK041322.1 | - | 2 | 212 | |||
| AK088021.1 | - | 2 | 159 | |||
| BC064058.1 | - | 2 | 139 | 3418931 | ||
| BY225764.1 | - | 2 | 227 | |||
| M34563.1 | - | 2 | 195 | |||

Cd28
Genomic Map