siRNA Details
| Gene Symbol: Scarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007644.3 | NP_031670.1 | 6, 7 | 1132 | ||
| GenBank mRNA | AB008553.1 | - | 6, 7 | 1037 | ||
| AK011123.1 | - | 6, 7 | 1062 | |||
| AK083038.1 | - | 6, 7 | 1133 | |||
| AK144235.1 | - | 6, 7 | 1164 | |||
| AK149102.1 | - | 1079 | ||||
| AK215493.1 | - | 1, 2 | 61 | |||
| AK215612.1 | - | 1, 2 | 50 | |||
| BC029073.1 | - | 6, 7 | 1040 | 3962669 | ||

Scarb2
Genomic Map