siRNA Details
| Gene Symbol: Cd3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013487.3 | NP_038515.3 | 2 | 253 | ||
| GenBank mRNA | AK042172.1 | - | 2 | 255 | ||
| AK050629.1 | - | 2 | 242 | |||
| AK153843.1 | - | 2 | 218 | |||
| AK154064.1 | - | 2 | 215 | |||
| BC062807.1 | - | 2 | 158 | 1366111 | ||
| EF382667.1 | - | 2 | 153 | |||
| HM536173.1 | - | 2 | 153 | |||
| X02339.1 | - | 2 | 121 | |||

Cd3d
Genomic Map