siRNA Details
| Gene Symbol: Cd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013488.2 | NP_038516.1 | 9 | 1459 | ||
| GenBank mRNA | AF045882.1 | - | 7 | 1218 | ||
| AK153966.1 | - | 9 | 1459 | |||
| AK161775.1 | - | 9 | 1383 | |||
| BC039137.1 | - | 9 | 1438 | 3670597 | ||
| M13816.1 | - | 8 | 1329 | |||
| M36850.1 | - | 8 | 1329 | |||
| U75219.1 | - | 3 | 244 | |||
| X04836.1 | - | 9 | 1459 | |||

Cd4
Genomic Map