siRNA Details
| Gene Symbol: Cd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009854.2 | NP_033984.1 | 2 | 213 | ||
| GenBank mRNA | AK136444.1 | - | 2 | 167 | ||
| AK154339.1 | - | 2 | 168 | |||
| D10329.1 | - | 2 | 153 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd7| Gene Symbol: Cd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009854.2 | NP_033984.1 | 2 | 213 | ||
| GenBank mRNA | AK136444.1 | - | 2 | 167 | ||
| AK154339.1 | - | 2 | 168 | |||
| D10329.1 | - | 2 | 153 | |||