siRNA Details
| Gene Symbol: Cd81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133655.2 | NP_598416.1 | 7 | 865 | ||
| GenBank mRNA | AK136370.1 | - | 7 | 829 | ||
| AK166521.1 | - | 7 | 831 | |||
| AK170741.1 | - | 7 | 830 | |||
| AK183391.1 | - | 2 | 81 | |||
| AK195226.1 | - | 81 | ||||
| BC011433.1 | - | 7 | 820 | 2811801 | ||
| X59047.1 | - | 7 | 585 | |||

Cd81
Genomic Map