siRNA Details
| Gene Symbol: Cd81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133655.2 | NP_598416.1 | 6, 7 | 820 | ||
| GenBank mRNA | AK136370.1 | - | 6, 7 | 784 | ||
| AK166521.1 | - | 6, 7 | 786 | |||
| AK170741.1 | - | 6, 7 | 785 | |||
| BC011433.1 | - | 6, 7 | 775 | 2811801 | ||
| X59047.1 | - | 6, 7 | 540 | |||

Cd81
Genomic Map