siRNA Details
| Gene Symbol: Cd81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133655.2 | NP_598416.1 | 7, 8 | 897 | ||
| GenBank mRNA | AK136370.1 | - | 7, 8 | 861 | ||
| AK166521.1 | - | 7, 8 | 863 | |||
| AK170741.1 | - | 7, 8 | 862 | |||
| AK195226.1 | - | 113 | ||||
| BC011433.1 | - | 7, 8 | 852 | 2811801 | ||
| X59047.1 | - | 7, 8 | 617 | |||

Cd81
Genomic Map