siRNA Details
| Gene Symbol: Cd8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009858.2 | NP_033988.1 | 2 | 282 | ||
| GenBank mRNA | AK087944.1 | - | 2 | 259 | ||
| AK088372.1 | - | 2 | 307 | |||
| AK088649.1 | - | 2 | 306 | |||
| AK154055.1 | - | 2 | 262 | |||
| AK162993.1 | - | 2 | 271 | |||
| AK169581.1 | - | 2 | 283 | |||
| M16799.1 | - | 2 | 275 | |||
| M17534.1 | - | 2 | 282 | |||
| M19504.1 | - | 2 | 270 | |||
| U34882.1 | - | 2 | 402 | |||
| X07698.1 | - | 2 | 286 | |||

Cd8b1
Genomic Map