siRNA Details
| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111075.4 | NP_001104545.1 | 7, 8 | 982 | ||
| NM_023117.4 | NP_075606.1 | 8, 9 | 1060 | |||
| GenBank mRNA | AK004836.1 | - | 7, 8 | 939 | ||
| AK043730.1 | - | 7, 8 | 973 | |||
| AK153903.1 | - | 8, 9 | 1061 | |||
| AK157397.1 | - | 7, 8 | 940 | |||
| BC048164.1 | - | 7, 8 | 979 | |||
| BC057568.1 | - | 7, 8 | 853 | 6401489 | ||
| S93521.1 | - | 8, 9 | 929 | |||

Cdc25b
Genomic Map