siRNA Details
| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111075.4 | NP_001104545.1 | 11 | 1370 | ||
| NM_023117.4 | NP_075606.1 | 12 | 1448 | |||
| GenBank mRNA | AK004836.1 | - | 11 | 1327 | ||
| AK043730.1 | - | 11 | 1361 | |||
| AK153903.1 | - | 12 | 1449 | |||
| BC002287.1 | - | 3 | 235 | |||
| BC048164.1 | - | 11 | 1367 | |||
| BC057568.1 | - | 11 | 1241 | 6401489 | ||
| S93521.1 | - | 12 | 1317 | |||

Cdc25b
Genomic Map