siRNA Details
| Gene Symbol: Cdk11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007661.3 | NP_031687.2 | 7 | 731 | ||
| GenBank mRNA | AK008283.1 | - | 7 | 708 | ||
| AK077668.1 | - | 7 | 732 | |||
| AK135402.1 | - | 7 | 738 | |||
| AK147133.1 | - | 7 | 709 | |||
| BC052920.1 | - | 7 | 809 | 6489832 | ||
| L37092.1 | - | 6 | 1306 | |||
| M58633.1 | - | 2 | 71 | |||

Cdk11b
Genomic Map