siRNA Details
| Gene Symbol: Cdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271566.1 | NP_001258495.1 | 6 | 843 | ||
| NM_001271567.1 | NP_001258496.1 | 6 | 674 | |||
| NM_001271568.1 | NP_001258497.1 | 6 | 674 | |||
| NM_009863.3 | NP_033993.2 | 6 | 674 | |||
| GenBank mRNA | AB018574.1 | - | 6 | 675 | ||
| AB018575.1 | - | 6 | 675 | |||
| AB018576.1 | - | 5 | 485 | |||
| AB018577.1 | - | 5 | 485 | |||
| AJ007661.1 | - | 6 | 626 | |||
| AK145921.1 | - | 6 | 600 | |||
| AK146009.1 | - | 6 | 845 | |||
| BC062149.1 | - | 6 | 604 | |||
| BC072666.1 | - | 6 | 615 | 30611704 | ||
| BC080702.1 | - | 6 | 627 | 30474846 | ||

Cdc7
Genomic Map