siRNA Details
| Gene Symbol: Cdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271566.1 | NP_001258495.1 | 9 | 1392 | ||
| NM_001271567.1 | NP_001258496.1 | 8 | 1127 | |||
| NM_001271568.1 | NP_001258497.1 | 8 | 1127 | |||
| NM_009863.3 | NP_033993.2 | 9 | 1223 | |||
| GenBank mRNA | AJ007661.1 | - | 9 | 1175 | ||
| AK145921.1 | - | 9 | 1149 | |||
| AK146009.1 | - | 9 | 1393 | |||
| BC062149.1 | - | 8 | 1941 | |||
| BC072666.1 | - | 9 | 1164 | 30611704 | ||
| BC080702.1 | - | 8 | 1080 | 30474846 | ||

Cdc7
Genomic Map