siRNA Details
| Gene Symbol: Arhgap31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020260.2 | NP_064656.2 | 12 | 3400 | ||
| GenBank mRNA | AK129309.1 | - | 3 | 1825 | ||
| AK149975.1 | - | 12 | 3236 | |||
| AK150109.1 | - | 12 | 3236 | |||
| AK155729.1 | - | 12 | 3220 | |||
| AK170685.1 | - | 12 | 3220 | |||
| BC138870.1 | - | 12 | 3205 | 8861892 | ||

Arhgap31
Genomic Map