siRNA Details
| Gene Symbol: Cdh17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019753.4 | NP_062727.1 | 13 | 1854 | ||
| GenBank mRNA | AF177669.1 | - | 13 | 1750 | ||
| AK155633.1 | - | 13 | 1855 | |||
| AK171733.1 | - | 13 | 1777 | |||
| BC042891.1 | - | 13 | 1772 | 4208360 | ||
| D87912.1 | - | 13 | 1799 | |||

Cdh17
Genomic Map