siRNA Details
| Gene Symbol: Cdh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009868.4 | NP_033998.2 | 10 | 1700 | ||
| GenBank mRNA | AK164474.1 | - | 10 | 1683 | ||
| AK167620.1 | - | 10 | 1632 | |||
| BC054790.1 | - | 10 | 1626 | 6831962 | ||
| D63942.1 | - | 10 | 1661 | |||
| X83930.2 | - | 10 | 1615 | |||

Cdh5
Genomic Map