siRNA Details
| Gene Symbol: Cdh9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009869.1 | NP_033999.1 | 2 | 298 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh9| Gene Symbol: Cdh9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009869.1 | NP_033999.1 | 2 | 298 | ||