siRNA Details
| Gene Symbol: Cdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016756.4 | NP_058036.1 | 6, 7 | 1010 | ||
| NM_183417.3 | NP_904326.1 | 7, 8 | 1154 | |||
| GenBank mRNA | AJ223732.1 | - | 7, 8 | 1056 | ||
| AK146845.1 | - | 7, 8 | 1134 | |||
| AK148017.1 | - | 7, 8 | 1093 | |||
| AK151979.1 | - | 7, 8 | 1104 | |||
| AK152922.1 | - | 6, 7 | 963 | |||
| AK153983.1 | - | 7, 8 | 1106 | |||
| AK154997.1 | - | 7, 8 | 1106 | |||
| AK178832.1 | - | 1 | 147 | |||
| AK179397.1 | - | 1, 2 | 147 | |||
| AK183240.1 | - | 1, 2 | 117 | |||
| BC005654.1 | - | 7, 8 | 1115 | 3709730 | ||
| BC026145.1 | - | 5, 6 | 1716 | |||
| U63337.1 | - | 6, 7 | 938 | |||

Cdk2
Genomic Map