siRNA Details
| Gene Symbol: Cdk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009873.2 | NP_034003.1 | 4 | 672 | ||
| GenBank mRNA | AF132482.1 | - | 4 | 503 | ||
| AF132483.1 | - | 4 | 672 | |||
| AK078973.1 | - | 4 | 783 | |||
| AK154197.1 | - | 5 | 945 | |||
| BC119360.1 | - | 4 | 661 | 8734109 | ||
| BC120595.1 | - | 4 | 661 | 40129518 | ||

Cdk6
Genomic Map