siRNA Details
| Gene Symbol: Cdkn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007670.4 | NP_031696.1 | 1, 2 | 368 | ||
| GenBank mRNA | AF059567.1 | - | 1, 2 | 277 | ||
| AK144132.1 | - | 1, 2 | 378 | |||
| AK146668.1 | - | 1, 2 | 329 | |||
| BC002010.1 | - | 1, 2 | 282 | 3495097 | ||
| DT919921.1 | - | 1, 2 | 107 | |||

Cdkn2b
Genomic Map