siRNA Details
| Gene Symbol: Cdkn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007670.4 | NP_031696.1 | 2 | 1286 | ||
| GenBank mRNA | AF059567.1 | - | 2 | 1194 | ||
| AK144132.1 | - | 2 | 1296 | |||
| BC002010.1 | - | 2 | 1199 | 3495097 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdkn2b| Gene Symbol: Cdkn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007670.4 | NP_031696.1 | 2 | 1286 | ||
| GenBank mRNA | AF059567.1 | - | 2 | 1194 | ||
| AK144132.1 | - | 2 | 1296 | |||
| BC002010.1 | - | 2 | 1199 | 3495097 | ||