siRNA Details
| Gene Symbol: Cdo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033037.3 | NP_149026.1 | 4 | 620 | ||
| GenBank mRNA | AK004249.1 | - | 4 | 634 | ||
| AK149582.1 | - | 4 | 634 | |||
| BC013638.1 | - | 4 | 620 | 4194939 | ||
| BC020375.1 | - | 4 | 620 | |||

Cdo1
Genomic Map