siRNA Details
| Gene Symbol: Mia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019394.3 | NP_062267.2 | 2 | 287 | ||
| GenBank mRNA | AK135151.1 | - | 2 | 266 | ||
| BC009815.1 | - | 2 | 256 | |||
| BE864464.1 | - | 2 | 287 | |||
| CK794645.1 | - | 2 | 281 | |||
| X94322.1 | - | 2 | 292 | |||

Mia
Genomic Map