siRNA Details
| Gene Symbol: Cdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007674.3 | NP_031700.1 | 3 | 892 | ||
| GenBank mRNA | AK141681.1 | - | 3 | 306 | ||
| BC119158.1 | - | 3 | 880 | 8733907 | ||
| L08061.1 | - | 3 | 895 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdx4| Gene Symbol: Cdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007674.3 | NP_031700.1 | 3 | 892 | ||
| GenBank mRNA | AK141681.1 | - | 3 | 306 | ||
| BC119158.1 | - | 3 | 880 | 8733907 | ||
| L08061.1 | - | 3 | 895 | |||