siRNA Details
Gene Symbol: Cdx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007674.3 | NP_031700.1 | 1 | 578 | ||
GenBank mRNA | BC119158.1 | - | 1 | 566 | 8733907 | |
L08061.1 | - | 1 | 581 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007674.3 | NP_031700.1 | 1 | 578 | ||
GenBank mRNA | BC119158.1 | - | 1 | 566 | 8733907 | |
L08061.1 | - | 1 | 581 |