siRNA Details
| Gene Symbol: Cdyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123386.1 | NP_001116858.1 | 2 | 499 | ||
| NM_009881.3 | NP_034011.1 | 5 | 449 | |||
| GenBank mRNA | AF081260.1 | - | 5 | 431 | ||
| AF081261.1 | - | 5 | 431 | |||
| AK028162.1 | - | 2 | 195 | |||
| AK156509.1 | - | 2 | 500 | |||
| BC055103.1 | - | 5 | 398 | 6741468 | ||
| BC062123.1 | - | 2 | 588 | 6826210 | ||

Cdyl
Genomic Map