siRNA Details
| Gene Symbol: Cer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009887.2 | NP_034017.1 | 2 | 768 | ||
| GenBank mRNA | AF031896.1 | - | 2 | 782 | ||
| AF035579.1 | - | 2 | 741 | |||
| BC132431.1 | - | 2 | 751 | 40130708 | ||
| BC145993.1 | - | 2 | 749 | 40131296 | ||

Cer1
Genomic Map