siRNA Details
| Gene Symbol: Cfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007685.2 | NP_031711.1 | 5, 6 | 754 | ||
| GenBank mRNA | BC100705.1 | - | 5, 6 | 712 | 40044593 | |
| BC100706.1 | - | 5, 6 | 712 | 40044594 | ||
| BC100708.1 | - | 5, 6 | 712 | 40044595 | ||
| BC100711.1 | - | 5, 6 | 712 | 40044596 | ||
| U57720.1 | - | 5, 6 | 754 | |||

Cfc1
Genomic Map