siRNA Details
| Gene Symbol: Cflar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289704.2 | NP_001276633.1 | 3 | 550 | ||
| NM_009805.4 | NP_033935.2 | 3 | 822 | |||
| NM_207653.5 | NP_997536.1 | 3 | 822 | |||
| GenBank mRNA | AK143812.1 | - | 3 | 815 | ||
| BC029223.1 | - | 3 | 791 | 4460554 | ||
| BC085105.1 | - | 3 | 545 | |||
| U97076.1 | - | 3 | 488 | |||
| Y14041.1 | - | 2 | 391 | |||
| Y14042.1 | - | 2 | 391 | |||

Cflar
Genomic Map