siRNA Details
| Gene Symbol: Cftr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021050.2 | NP_066388.1 | 17 | 2941 | ||
| GenBank mRNA | M60493.1 | - | 17 | 2942 | ||
| M69298.1 | - | 17 | 2940 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cftr| Gene Symbol: Cftr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021050.2 | NP_066388.1 | 17 | 2941 | ||
| GenBank mRNA | M60493.1 | - | 17 | 2942 | ||
| M69298.1 | - | 17 | 2940 | |||