siRNA Details
| Gene Symbol: Cftr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021050.2 | NP_066388.1 | 8 | 1212 | ||
| GenBank mRNA | AK033621.1 | - | 8 | 1130 | ||
| M60493.1 | - | 8 | 1213 | |||
| M69298.1 | - | 8 | 1211 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cftr| Gene Symbol: Cftr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021050.2 | NP_066388.1 | 8 | 1212 | ||
| GenBank mRNA | AK033621.1 | - | 8 | 1130 | ||
| M60493.1 | - | 8 | 1213 | |||
| M69298.1 | - | 8 | 1211 | |||