siRNA Details
| Gene Symbol: Chrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278041.1 | NP_001264970.1 | 6 | 721 | ||
| NM_009893.2 | NP_034023.1 | 6 | 655 | |||
| GenBank mRNA | AF069501.1 | - | 6 | 655 | ||
| AF096276.1 | - | 6 | 634 | |||
| AK077460.1 | - | 6 | 651 | |||
| AK149445.1 | - | 6 | 630 | |||
| AK165339.1 | - | 6 | 648 | |||
| BC145017.1 | - | 6 | 719 | |||
| BC145018.1 | - | 6 | 719 | |||
| BC145019.1 | - | 6 | 653 | |||
| BC145696.1 | - | 6 | 719 | 40130999 | ||

Chrd
Genomic Map