siRNA Details
| Gene Symbol: Chrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278041.1 | NP_001264970.1 | 22 | 2856 | ||
| NM_009893.2 | NP_034023.1 | 22 | 2790 | |||
| GenBank mRNA | AF069501.1 | - | 22 | 2790 | ||
| AF096276.1 | - | 22 | 2769 | |||
| AK077460.1 | - | 23 | 2954 | |||
| AK149445.1 | - | 22 | 2765 | |||
| AK220310.1 | - | 5 | 3594 | |||
| BC145017.1 | - | 20 | 2904 | |||
| BC145018.1 | - | 22 | 2824 | |||
| BC145019.1 | - | 22 | 2861 | |||
| BC145696.1 | - | 22 | 2854 | 40130999 | ||

Chrd
Genomic Map