siRNA Details
| Gene Symbol: Chrm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033269.4 | NP_150372.1 | 5 | 1407 | ||
| GenBank mRNA | AK080950.1 | - | 5 | 1409 | ||
| BC129892.1 | - | 5 | 902 | 40111498 | ||
| BC129893.1 | - | 5 | 902 | 40111499 | ||
More Information
Other Species Targeted
Set Membership: |
|

Chrm3
Genomic Map