siRNA Details
| Gene Symbol: Cirbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007705.2 | NP_031731.1 | 2 | 174 | ||
| GenBank mRNA | AK014655.1 | - | 2 | 177 | ||
| AK132584.1 | - | 2 | 177 | |||
| BC075699.1 | - | 2 | 164 | 30656047 | ||
| D78135.1 | - | 2 | 174 | |||

Cirbp
Genomic Map